Assay Details
Target Gene Details
Entrez Gene ID: | 23272 |
Gene Name: | family with sequence similarity 208 member A |
Gene Aliases: |
C3orf63, RAP140, se89-1 |
Location: |
Chr.3:56620132-56683242 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM208A | NM_001112736.1 | 18 | 3412 | NP_001106207.1 |
| NM_015224.3 | 11 | 2640 | NP_056039.2 | |
| XM_005264999.1 | 18 | 3547 | XP_005265056.1 | |
| XM_006713077.2 | 18 | 3424 | XP_006713140.1 | |
| XM_006713078.2 | 12 | 2401 | XP_006713141.1 | |
| XM_011533552.2 | 17 | 3364 | XP_011531854.1 | |
| XM_011533553.2 | 17 | 2968 | XP_011531855.1 | |
| XM_017006030.1 | 18 | 3199 | XP_016861519.1 | |
| XM_017006031.1 | 17 | 3364 | XP_016861520.1 | |
| XM_017006032.1 | 17 | 2845 | XP_016861521.1 | |
| AB029028.1 | 1 | 493 | BAA83057.1 | |
| AF180425.2 | 11 | 2924 | AAD55098.1 | |
| AF273053.1 | 4 | 1564 | AAG34913.1 | |
| AK307599.1 | 10 | 2102 | ||
| BC129987.1 | 10 | 2188 | ||
| BX648955.1 | 17 | 2894 |
More Information
Set Membership: |
|

Genomic Map