Assay Details
Target Gene Details
Entrez Gene ID: | 399664 |
Gene Name: | mex-3 RNA binding family member D |
Gene Aliases: |
MEX-3D, MEX3, OK/SW-cl.4, RKHD1, RNF193, TINO |
Location: |
Chr.19:1554669-1568058 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEX3D | NM_001174118.1 | 2 | 745 | NP_001167589.1 |
| NM_203304.3 | 2 | 745 | NP_976049.3 | |
| XM_017026811.1 | 2 | 545 | XP_016882300.1 | |
| AB107353.1 | 2 | 338 | BAC67658.1 | |
| AF458084.1 | 2 | 217 | AAQ04763.1 | |
| AY950680.1 | 2 | 745 | ||
| BC113384.1 | 2 | 216 | ||
| BC113741.1 | 2 | 216 | ||
| DQ225272.1 | 3 | 415 | ABB04031.1 | |
| DQ225273.1 | ABB04032.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss |
|
| nsv517758 | Chr.19:1550651 - 1572370 on Build GRCh38 | Loss |
|
| nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map