Assay Details
Target Gene Details
Entrez Gene ID: | 54478 |
Gene Name: | family with sequence similarity 64 member A |
Gene Aliases: |
CATS, RCS1 |
Location: |
Chr.17:6444305-6451065 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM64A | NM_001195228.1 | NP_001182157.1 | ||
| NM_019013.2 | NP_061886.2 | |||
| XM_017024778.1 | XP_016880267.1 | |||
| AK001018.1 | BAA91468.1 | |||
| AK001353.1 | BAA91644.1 | |||
| AK225702.1 | ||||
| BC005004.1 | AAH05004.1 | |||
| BC013966.2 | AAH13966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574278 | Chr.17:5546345 - 6722066 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map