Assay Details
Target Gene Details
Entrez Gene ID: | 196374 |
Gene Name: | keratin 78 |
Gene Aliases: |
CK-78, K5B, K78, Kb40 |
Location: |
Chr.12:52837804-52848994 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT78 | NM_173352.3 | NP_775487.2 | ||
| XM_011538010.1 | XP_011536312.1 | |||
| AK096419.1 | BAC04782.1 | |||
| AK290901.1 | ||||
| AK298636.1 | ||||
| DA667429.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv711 | Chr.12:52841102 - 52873555 on Build GRCh38 | Insertion |
|
| nsv832416 | Chr.12:52779656 - 52971582 on Build GRCh38 | Loss |
|
| nsv1048816 | Chr.12:52811390 - 52861371 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116117459] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map