Assay Details
Target Gene Details
Entrez Gene ID: | 6622 |
Gene Name: | synuclein alpha |
Gene Aliases: |
NACP, PARK1, PARK4, PD1 |
Location: |
Chr.4:89724099-89838324 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNCA | NM_000345.3 | NP_000336.1 | ||
| NM_001146054.1 | NP_001139526.1 | |||
| NM_001146055.1 | NP_001139527.1 | |||
| NM_007308.2 | NP_009292.1 | |||
| XM_011532203.1 | XP_011530505.1 | |||
| XM_011532204.2 | 1 | 128 | XP_011530506.1 | |
| XM_011532205.2 | XP_011530507.1 | |||
| XM_011532206.1 | XP_011530508.1 | |||
| XM_011532208.2 | XP_011530510.1 | |||
| AI815523.1 | ||||
| AK290169.1 | ||||
| BC013293.2 | 1 | 154 | AAH13293.1 | |
| BC108275.1 | AAI08276.1 | |||
| CR736451.1 | ||||
| DA391115.1 | ||||
| HQ830267.1 | ||||
| HQ830268.1 | 1 | 166 | ||
| HQ830269.1 | ||||
| JN709859.1 | ||||
| JN709860.1 | ||||
| JN709861.1 | ||||
| JN709862.1 | ||||
| JN709863.1 | ||||
| JN709864.1 | ||||
| JN709865.1 | ||||
| JN709866.1 | ||||
| L08850.1 | AAA16117.1 |
Target Gene Details
Entrez Gene ID: | 644248 |
Gene Name: | SNCA antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:89836401-89841991 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNCA-AS1 | NR_045481.1 | |||
| AK123890.1 | ||||
| BG181213.1 | ||||
| KF728371.1 | 1 | 86 | ||
| KF728372.1 | 1 | 86 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012406 | Chr.4:89246630 - 90245636 on Build GRCh38 | Loss |
|
| nsv594806 | Chr.4:88621130 - 90539537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map