Assay Details
Target Gene Details
Entrez Gene ID: | 57569 |
Gene Name: | Rho GTPase activating protein 20 |
Gene Aliases: |
RARHOGAP |
Location: |
Chr.11:110577035-110713189 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP20 | NM_001258415.1 | 15 | 3762 | NP_001245344.1 |
| NM_001258416.1 | 16 | 3648 | NP_001245345.1 | |
| NM_001258417.1 | 15 | 3462 | NP_001245346.1 | |
| NM_001258418.1 | 15 | 3580 | NP_001245347.1 | |
| NM_020809.3 | 16 | 3759 | NP_065860.2 | |
| XM_005271628.3 | 5 | 2433 | XP_005271685.1 | |
| XM_006718893.3 | 15 | 3668 | XP_006718956.1 | |
| AB037812.1 | 15 | 3484 | ||
| AK292290.1 | 5 | 2433 | ||
| AY496263.1 | 16 | 3759 | AAS45466.1 | |
| AY496264.1 | 15 | 3462 | AAS45467.1 | |
| AY496265.1 | 15 | 3580 | AAS45468.1 | |
| AY496266.1 | 15 | 3762 | AAS45469.1 | |
| AY496267.1 | 16 | 3648 | AAS45470.1 | |
| BC039340.1 | 5 | 2418 | AAH39340.1 |
More Information
Set Membership: |
|

Genomic Map