Assay Details
Target Gene Details
Entrez Gene ID: | 553103 |
Gene Name: | uncharacterized LOC553103 |
Gene Aliases: |
- |
Location: |
Chr.5:132311276-132369916 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC553103 | NR_110997.1 |
Target Gene Details
Entrez Gene ID: | 6583 |
Gene Name: | solute carrier family 22 member 4 |
Gene Aliases: |
OCTN1 |
Location: |
Chr.5:132294384-132344206 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC22A4 | NM_003059.2 | 7 | 1252 | NP_003050.2 |
| XM_006714675.3 | 6 | 659 | XP_006714738.1 | |
| XM_011543589.2 | 5 | 1044 | XP_011541891.1 | |
| XM_017009776.1 | 7 | 1300 | XP_016865265.1 | |
| AB007448.1 | 7 | 1233 | BAA23356.1 | |
| BC028313.1 | 7 | 1252 | AAH28313.1 | |
| Y09881.1 | 7 | 1245 | CAA71007.1 |
More Information
Set Membership: |
|

Genomic Map