Assay Details
Target Gene Details
Entrez Gene ID: | 90522 |
Gene Name: | Yip1 interacting factor homolog B, membrane trafficking protein |
Gene Aliases: |
FinGER8 |
Location: |
Chr.19:38303560-38315976 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YIF1B | NM_001039671.2 | 8 | 1201 | NP_001034760.1 |
| NM_001039672.2 | 8 | 1219 | NP_001034761.1 | |
| NM_001039673.2 | 8 | 1210 | NP_001034762.1 | |
| NM_001145461.1 | 8 | 1195 | NP_001138933.1 | |
| NM_001145462.1 | 8 | 1171 | NP_001138934.1 | |
| NM_001145463.1 | 9 | 1287 | NP_001138935.1 | |
| XM_005259385.3 | XP_005259442.1 | |||
| XM_017027450.1 | XP_016882939.1 | |||
| XM_017027451.1 | XP_016882940.1 | |||
| AF258578.3 | AAG23781.1 | |||
| AL833382.1 | CAI46138.1 | |||
| AY358965.1 | AAQ89324.1 | |||
| BC007644.1 | AAH07644.1 | |||
| BC091477.1 | AAH91477.2 | |||
| C02248.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833822 | Chr.19:38270108 - 38454465 on Build GRCh38 | Loss |
|

Genomic Map