Assay Details
Target Gene Details
Entrez Gene ID: | 8493 |
Gene Name: | protein phosphatase, Mg2+/Mn2+ dependent 1D |
Gene Aliases: |
PP2C-DELTA, WIP1 |
Location: |
Chr.17:60600183-60666280 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPM1D | NM_003620.3 | 2 | 891 | NP_003611.1 |
| AK299773.1 | 3 | 600 | ||
| AK312902.1 | 2 | 881 | ||
| BC016480.1 | 2 | 807 | AAH16480.1 | |
| BC033893.2 | 2 | 765 | AAH33893.1 | |
| BC042418.1 | 2 | 805 | AAH42418.1 | |
| BC060877.1 | 2 | 891 | AAH60877.1 | |
| BT009780.1 | 2 | 659 | AAP88782.1 | |
| KU178420.1 | 2 | 659 | ||
| KU178421.1 | ||||
| U78305.1 | 2 | 862 | AAB61637.1 |
More Information
Set Membership: |
|

Genomic Map