Assay Details
Target Gene Details
Entrez Gene ID: | 10300 |
Gene Name: | katanin regulatory subunit B1 |
Gene Aliases: |
KAT, LIS6 |
Location: |
Chr.16:57735717-57757250 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 17 - Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KATNB1 | NM_005886.2 | NP_005877.2 | ||
| XM_005255772.4 | XP_005255829.1 | |||
| XM_006721121.3 | XP_006721184.1 | |||
| XM_006721122.2 | XP_006721185.1 | |||
| XM_006721123.3 | XP_006721186.1 | |||
| XM_011522810.2 | XP_011521112.1 | |||
| XM_017022860.1 | XP_016878349.1 | |||
| XM_017022861.1 | XP_016878350.1 | |||
| XM_017022862.1 | XP_016878351.1 | |||
| XM_017022863.1 | XP_016878352.1 | |||
| XM_017022864.1 | XP_016878353.1 | |||
| AB209250.1 | BAD92487.1 | |||
| AF052432.1 | AAC09328.1 | |||
| BC001353.2 | AAH01353.1 | |||
| BT007022.1 | AAP35668.1 | |||
| CA306372.1 | ||||
| CR456762.1 | CAG33043.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523237 | Chr.16:57753115 - 57786197 on Build GRCh38 | Loss |
|
| nsv471089 | Chr.16:57722731 - 57788355 on Build GRCh38 | Loss |
|
| nsv525464 | Chr.16:57714251 - 57832787 on Build GRCh38 | Gain |
|
| nsv509622 | Chr.16:57720104 - 57826484 on Build GRCh38 | Insertion |
|
| nsv528250 | Chr.16:57715692 - 57786197 on Build GRCh38 | Loss |
|
| nsv952038 | Chr.16:57734189 - 57774488 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map