Assay Details
Target Gene Details
Entrez Gene ID: | 57613 |
Gene Name: | family with sequence similarity 234 member B |
Gene Aliases: |
KIAA1467 |
Location: |
Chr.12:12927680-13083449 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM234B | XM_017019706.1 | XP_016875195.1 |
Target Gene Details
Entrez Gene ID: | 55507 |
Gene Name: | G protein-coupled receptor class C group 5 member D |
Gene Aliases: |
- |
Location: |
Chr.12:12940518-12955908 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPRC5D | NM_018654.1 | NP_061124.1 | ||
| XM_005253421.3 | XP_005253478.1 | |||
| XM_017019582.1 | XP_016875071.1 | |||
| XM_017019583.1 | XP_016875072.1 | |||
| AF209923.1 | AAF72873.1 | |||
| BC069341.1 | AAH69341.1 | |||
| BC107077.1 | AAI07078.1 | |||
| BC107078.1 | AAI07079.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion |
|
| nsv832333 | Chr.12:12896803 - 13108130 on Build GRCh38 | Gain |
|
| nsv520173 | Chr.12:12930203 - 12972003 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map