Assay Details
Target Gene Details
Entrez Gene ID: | 3232 |
Gene Name: | homeobox D3 |
Gene Aliases: |
HOX1D, HOX4, HOX4A, Hox-4.1 |
Location: |
Chr.2:176136921-176173102 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXD3 | XM_005246510.4 | XP_005246567.1 | ||
| XM_006712477.2 | XP_006712540.1 |
Target Gene Details
Entrez Gene ID: | 3233 |
Gene Name: | homeobox D4 |
Gene Aliases: |
HHO.C13, HOX-5.1, HOX4, HOX4B, Hox-4.2 |
Location: |
Chr.2:176150394-176156078 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXD4 | NM_014621.2 | NP_055436.2 | ||
| XM_005246514.4 | XP_005246571.1 | |||
| AK313885.1 | ||||
| BC016763.1 | AAH16763.1 | |||
| BC074797.2 | AAH74797.1 | |||
| X04706.1 | CAA28411.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv676n67 | Chr.2:176057192 - 176216223 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map