Assay Details
Target Gene Details
Entrez Gene ID: | 60312 |
Gene Name: | actin filament associated protein 1 |
Gene Aliases: |
AFAP, AFAP-110, AFAP110 |
Location: |
Chr.4:7758713-7939926 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AFAP1 | NM_001134647.1 | 18 | 4223 | NP_001128119.1 |
| NM_198595.2 | 17 | 3971 | NP_940997.1 | |
| XM_006713909.3 | 17 | 3854 | XP_006713972.1 | |
| XM_011513544.2 | 18 | 4276 | XP_011511846.1 | |
| XM_017008535.1 | 17 | 3754 | XP_016864024.1 | |
| AF188700.1 | 16 | 3698 | AAG17055.1 | |
| D25248.1 | 2 | 1548 |
Target Gene Details
Entrez Gene ID: | 84740 |
Gene Name: | AFAP1 antisense RNA 1 |
Gene Aliases: |
AFAP1-AS, AFAP1AS |
Location: |
Chr.4:7754090-7778928 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AFAP1-AS1 | NR_026892.1 |
More Information
Set Membership: |
|

Genomic Map