Assay Details
Target Gene Details
Entrez Gene ID: | 2802 |
Gene Name: | golgin A3 |
Gene Aliases: |
GCP170, MEA-2 |
Location: |
Chr.12:132768909-132829131 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLGA3 | NM_005895.3 | 24 | 8791 | NP_005886.2 |
| XM_005266164.4 | 24 | 8398 | XP_005266221.1 | |
| XM_005266167.4 | 21 | 7823 | XP_005266224.1 | |
| XM_006719736.3 | 24 | 8518 | XP_006719799.1 | |
| XM_006719737.3 | 24 | 8528 | XP_006719800.1 | |
| XM_011534793.2 | 23 | 8245 | XP_011533095.1 | |
| XM_011534794.2 | 16 | 6396 | XP_011533096.1 | |
| AK092223.1 | 1 | 1568 | ||
| BM981309.1 | 1 | 480 | ||
| D63997.1 | 25 | 6183 | BAA23661.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3631312 | Chr.12:132735138 - 132794488 on Build GRCh38 | Loss |
|
| esv3631315 | Chr.12:132764786 - 132771466 on Build GRCh38 | Loss |
|
| nsv561129 | Chr.12:132736267 - 132769626 on Build GRCh38 | Loss |
|
| esv3580555 | Chr.12:132760405 - 132796737 on Build GRCh38 | Loss |
|
| nsv1037074 | Chr.12:132613063 - 132801953 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map