Assay Details
Target Gene Details
Entrez Gene ID: | 3742 |
Gene Name: | potassium voltage-gated channel subfamily A member 6 |
Gene Aliases: |
HBK2, KV1.6, PPP1R96 |
Location: |
Chr.12:4806877-4851927 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNA6 | NM_002235.3 | 1 | 463 | NP_002226.1 |
| XM_005253686.3 | 1 | 2762 | XP_005253743.1 | |
| XM_011520955.2 | 1 | 2762 | XP_011519257.1 | |
| XM_017019270.1 | 1 | 2762 | XP_016874759.1 | |
| XM_017019271.1 | 1 | 2762 | XP_016874760.1 | |
| XM_017019272.1 | 1 | 2762 | XP_016874761.1 | |
| X17622.1 | 1 | 463 | CAA35623.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483060 | Chr.12:3190835 - 5290834 on Build GRCh38 | Loss |
|
| nsv1040098 | Chr.12:3987637 - 5646161 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map