Assay Details
Target Gene Details
Entrez Gene ID: | 8861 |
Gene Name: | LIM domain binding 1 |
Gene Aliases: |
CLIM-2, CLIM2, LDB-1, NLI |
Location: |
Chr.10:102102100-102120473 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDB1 | NM_001113407.2 | NP_001106878.1 | ||
| NM_001321612.1 | NP_001308541.1 | |||
| NM_003893.4 | NP_003884.1 | |||
| XM_017016867.1 | XP_016872356.1 | |||
| XM_017016868.1 | XP_016872357.1 | |||
| XM_017016869.1 | XP_016872358.1 | |||
| AB016485.1 | BAA31991.1 | |||
| AB250384.1 | ||||
| AF064491.1 | AAC28341.1 | |||
| AF068652.1 | AAC77818.1 | |||
| AK300588.1 | ||||
| AK308481.1 | ||||
| BC000482.2 | AAH00482.1 | |||
| BC009246.2 | AAH09246.1 | |||
| BT007054.1 | AAP35703.1 |
More Information
Set Membership: |
|

Genomic Map