Assay Details
Target Gene Details
Entrez Gene ID: | 10169 |
Gene Name: | small EDRK-rich factor 2 |
Gene Aliases: |
4F5REL, FAM2C, H4F5REL, HsT17089 |
Location: |
Chr.15:43777096-43796089 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERF2 | NM_001018108.3 | 3 | 2383 | NP_001018118.1 |
| NM_001199875.1 | 3 | 2383 | NP_001186804.1 | |
| NM_001199876.1 | 3 | 2272 | NP_001186805.1 | |
| NM_001199877.1 | 5 | 2781 | NP_001186806.1 | |
| NM_001199878.1 | 3 | 2076 | NP_001186807.1 | |
| NR_037672.1 | 4 | 1812 | ||
| AK094846.1 | 1 | 1065 | ||
| BM970646.1 |
Target Gene Details
Entrez Gene ID: | 100529067 |
Gene Name: | SERF2-C15orf63 readthrough |
Gene Aliases: |
- |
Location: |
Chr.15:43791976-43802589 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERF2-C15ORF63 | NR_037673.1 | |||
| AK000438.1 | BAA91165.1 | |||
| AK095876.1 |
Target Gene Details
Entrez Gene ID: | 619189 |
Gene Name: | serine incorporator 4 |
Gene Aliases: |
- |
Location: |
Chr.15:43794174-43800097 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERINC4 | NM_001258031.1 | NP_001244960.1 | ||
| NM_001258032.1 | NP_001244961.1 | |||
| AK097682.1 | ||||
| BC136669.1 | ||||
| BX647438.1 | ||||
| DQ103711.1 | AAZ80298.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1036318 | Chr.15:43761419 - 43868575 on Build GRCh38 | Gain |
|
| esv2760028 | Chr.15:43530960 - 43856443 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map