Assay Details
Target Gene Details
Entrez Gene ID: | 8354 |
Gene Name: | histone cluster 1, H3i |
Gene Aliases: |
H3.f, H3/f, H3FF |
Location: |
Chr.6:27871845-27872321 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIST1H3I | NM_003533.2 | NP_003524.1 | ||
| BC066884.1 | AAH66884.1 | |||
| BC067493.1 | AAH67493.1 | |||
| BC069305.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv601208 | Chr.6:27842848 - 28001226 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map