Assay Details
Target Gene Details
Entrez Gene ID: | 80760 |
Gene Name: | inter-alpha-trypsin inhibitor heavy chain family member 5 |
Gene Aliases: |
ITI-HC5, PP14776 |
Location: |
Chr.10:7559270-7666998 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITIH5 | NM_030569.6 | 13 | 2358 | NP_085046.5 |
| NM_032817.5 | 9 | 1693 | NP_116206.4 | |
| XM_011519713.2 | 14 | 2433 | XP_011518015.1 | |
| AB075833.1 | 10 | 1899 | BAB85539.1 | |
| AK027375.1 | 13 | 2295 | BAB55070.1 | |
| AK075381.1 | 13 | 2331 | ||
| AL833203.1 | 13 | 2203 | ||
| AY238437.1 | 13 | 2254 | AAO49812.1 | |
| AY358426.1 | AAQ88792.1 | |||
| BE898209.1 | 1 | 79 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038398 | Chr.10:7501987 - 7614812 on Build GRCh38 | Gain |
|
| nsv1038131 | Chr.10:7416409 - 7706490 on Build GRCh38 | Gain |
|
| esv2759732 | Chr.10:7381534 - 7695776 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map