Assay Details
Target Gene Details
Entrez Gene ID: | 8658 |
Gene Name: | tankyrase |
Gene Aliases: |
ARTD5, PARP-5a, PARP5A, PARPL, TIN1, TINF1, TNKS1, pART5 |
Location: |
Chr.8:9555229-9782346 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNKS | NM_003747.2 | NP_003738.2 | ||
| XM_006716263.3 | 1 | 733 | XP_006716326.1 | |
| XM_011543845.2 | 1 | 733 | XP_011542147.1 | |
| XM_011543846.2 | 1 | 733 | XP_011542148.1 | |
| XM_011543847.2 | 1 | 732 | XP_011542149.1 | |
| AF082556.1 | AAC79841.1 | |||
| AK302031.1 | ||||
| BC098394.1 | AAH98394.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1033031 | Chr.8:9452756 - 9561431 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map