Assay Details
Target Gene Details
Entrez Gene ID: | 338785 |
Gene Name: | keratin 79 |
Gene Aliases: |
K6L, KRT6L |
Location: |
Chr.12:52821447-52834293 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT79 | NM_175834.2 | 2 | 648 | NP_787028.1 |
| AJ564105.1 | 2 | 633 | CAD91893.1 | |
| BC039148.1 | 2 | 650 | AAH39148.1 | |
| BC063648.1 | 2 | 647 | AAH63648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832416 | Chr.12:52779656 - 52971582 on Build GRCh38 | Loss |
|
| nsv1048816 | Chr.12:52811390 - 52861371 on Build GRCh38 | Gain |
|
| esv3629524 | Chr.12:52688070 - 52835680 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map