Assay Details
Target Gene Details
Entrez Gene ID: | 63910 |
Gene Name: | solute carrier family 17 member 9 |
Gene Aliases: |
C20orf59, POROK8, VNUT |
Location: |
Chr.20:62952647-62968597 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC17A9 | NM_001302643.1 | 1 | 98 | NP_001289572.1 |
| NM_022082.3 | 1 | 98 | NP_071365.3 | |
| XM_011528978.2 | XP_011527280.1 | |||
| AK074107.1 | BAB84933.1 | |||
| AK125597.1 | ||||
| DB153870.1 | 1 | 98 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470559 | Chr.20:62945923 - 62974966 on Build GRCh38 | Loss |
|
| nsv586532 | Chr.20:62929801 - 62968122 on Build GRCh38 | Gain |
|
| nsv9825 | Chr.20:62775470 - 63601330 on Build GRCh38 | Gain |
|
| dgv7659n54 | Chr.20:62951856 - 62968122 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map