Assay Details
Target Gene Details
Entrez Gene ID: | 4222 |
Gene Name: | mesenchyme homeobox 1 |
Gene Aliases: |
KFS2, MOX1 |
Location: |
Chr.17:43640389-43661954 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEOX1 | NM_001040002.1 | 4 | 1766 | NP_001035091.1 |
| NM_004527.3 | 3 | 1912 | NP_004518.1 | |
| NM_013999.3 | 2 | 1739 | NP_054705.1 | |
| BG677020.1 | 1 | 723 | ||
| BQ003431.1 | 1 | 409 | ||
| BQ028272.1 | 1 | 419 | ||
| U10492.1 | 3 | 1912 | AAA19983.1 | |
| U10493.1 | 2 | 1739 | AAA19984.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060861 | Chr.17:43338154 - 43791949 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map