Assay Details
Target Gene Details
Entrez Gene ID: | 25875 |
Gene Name: | LETM1 domain containing 1 |
Gene Aliases: |
1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2 |
Location: |
Chr.12:51048230-51068984 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3629498 | Chr.12:51040936 - 51066446 on Build GRCh38 | Gain |
|
| esv34039 | Chr.12:50932919 - 51278127 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map