Assay Details
Target Gene Details
Entrez Gene ID: | 84173 |
Gene Name: | ELMO domain containing 3 |
Gene Aliases: |
DFNB88, LST3, RBED1, RBM29 |
Location: |
Chr.2:85354394-85391752 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELMOD3 | NM_001135021.1 | 5 | 587 | NP_001128493.1 |
| NM_001135022.1 | 4 | 403 | NP_001128494.1 | |
| NM_001135023.1 | 3 | 359 | NP_001128495.1 | |
| NM_032213.4 | 2 | 264 | NP_115589.2 | |
| XM_005264596.3 | 4 | 403 | XP_005264653.1 | |
| XM_005264597.3 | 3 | 685 | XP_005264654.1 | |
| XM_005264598.3 | 3 | 308 | XP_005264655.1 | |
| XM_017005078.1 | 3 | 259 | XP_016860567.1 | |
| XM_017005079.1 | 2 | 215 | XP_016860568.1 | |
| AF258573.1 | AAG23776.1 | |||
| AK307252.1 | 3 | 377 | ||
| BC001942.1 | 2 | 181 | AAH01942.1 | |
| BC010991.2 | 4 | 335 | AAH10991.1 | |
| BC018666.2 | 2 | 181 | AAH18666.1 | |
| BC112324.1 | 3 | 322 | AAI12325.1 | |
| DA294897.1 | 3 | 312 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575266 | Chr.2:85065061 - 85379120 on Build GRCh38 | Gain |
|
| nsv834282 | Chr.2:85237271 - 85414189 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map