Assay Details
Target Gene Details
Entrez Gene ID: | 144455 |
Gene Name: | E2F transcription factor 7 |
Gene Aliases: |
- |
Location: |
Chr.12:77021246-77065580 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E2F7 | NM_203394.2 | 13 | 5025 | NP_976328.2 |
| XM_011537966.2 | 12 | 4890 | XP_011536268.1 | |
| XM_011537969.2 | 12 | 4588 | XP_011536271.1 | |
| AK023043.1 | 1 | 788 | ||
| AK096316.1 | 5 | 3234 | ||
| BC016658.1 | 3 | 2584 | AAH16658.1 |
More Information
Set Membership: |
|

Genomic Map