Assay Details
Target Gene Details
Entrez Gene ID: | 79861 |
Gene Name: | tubulin alpha like 3 |
Gene Aliases: |
- |
Location: |
Chr.10:5393098-5404830 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBAL3 | NM_001171864.1 | NP_001165335.1 | ||
| NM_024803.2 | NP_079079.1 | |||
| AK025318.1 | BAB15110.1 | |||
| AK296616.1 | ||||
| BC098247.1 | AAH98247.1 | |||
| BC099716.1 | AAH99716.1 | |||
| BC105634.1 | AAI05635.1 | |||
| DA890426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549886 | Chr.10:5163474 - 5590097 on Build GRCh38 | Loss |
|
| nsv1045141 | Chr.10:5384177 - 5449996 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map