Assay Details
Target Gene Details
Entrez Gene ID: | 57402 |
Gene Name: | S100 calcium binding protein A14 |
Gene Aliases: |
BCMP84, S100A15 |
Location: |
Chr.1:153614255-153616917 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A14 | NM_020672.2 | 4 | 738 | NP_065723.1 |
| XM_005245362.1 | 4 | 734 | XP_005245419.1 | |
| XM_017001875.1 | 5 | 826 | XP_016857364.1 | |
| AY007220.1 | 4 | 720 | AAG01893.1 | |
| BG677973.1 | 4 | 738 |
More Information
Set Membership: |
|

Genomic Map