Assay Details
Target Gene Details
Entrez Gene ID: | 5551 |
Gene Name: | perforin 1 |
Gene Aliases: |
FLH2, HPLH2, P1, PFN1, PFP |
Location: |
Chr.10:70597348-70602775 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRF1 | NM_001083116.1 | NP_001076585.1 | ||
| NM_005041.4 | NP_005032.2 | |||
| BC047695.1 | ||||
| BC063043.1 | AAH63043.1 | |||
| BQ654088.1 | ||||
| FJ555237.1 | ||||
| L40557.1 | AAA63618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss |
|
| nsv1043245 | Chr.10:70290210 - 70915897 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map