Assay Details
Target Gene Details
Entrez Gene ID: | 8192 |
Gene Name: | caseinolytic mitochondrial matrix peptidase proteolytic subunit |
Gene Aliases: |
DFNB81, PRLTS3 |
Location: |
Chr.19:6361452-6368904 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLPP | NM_006012.2 | 1 | 89 | NP_006003.1 |
| AK311973.1 | ||||
| BC002956.1 | 1 | 89 | AAH02956.1 | |
| EF036491.1 | ||||
| Z50853.1 | CAA90705.1 |
More Information
Set Membership: |
|

Genomic Map