Assay Details
Target Gene Details
Entrez Gene ID: | 83440 |
Gene Name: | ADP dependent glucokinase |
Gene Aliases: |
2610017G09Rik, ADP-GK |
Location: |
Chr.15:72751367-72783785 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADPGK | NM_031284.4 | 7 | 2258 | NP_112574.3 |
| NR_023318.1 | 7 | 2471 | ||
| NR_023319.1 | 5 | 1972 | ||
| XM_006720712.3 | 7 | 2261 | XP_006720775.1 | |
| XM_006720713.3 | 7 | 2422 | XP_006720776.1 | |
| XM_011522102.2 | 4 | 1844 | XP_011520404.1 | |
| XM_011522103.2 | 4 | 1841 | XP_011520405.1 | |
| XM_017022664.1 | 3 | 2202 | XP_016878153.1 | |
| AF204270.1 | 6 | 1850 | AAL31473.1 | |
| AK055526.1 | 5 | 1963 | BAB70941.1 | |
| AK057651.1 | 2 | 2001 | ||
| AK075560.1 | 7 | 2193 | BAC11699.1 | |
| AK095995.1 | 2 | 2350 | ||
| AK293282.1 | 9 | 1554 | ||
| AL136873.1 | 4 | 1521 | CAB66807.1 | |
| AL832254.1 | 2 | 4279 | CAH10598.1 | |
| BE018185.1 | 1 | 142 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760039 | Chr.15:72527444 - 72828219 on Build GRCh38 | Gain+Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map