Assay Details
Target Gene Details
Entrez Gene ID: | 6871 |
Gene Name: | transcriptional adaptor 2A |
Gene Aliases: |
ADA2, ADA2A, KL04P, TADA2L, hADA2 |
Location: |
Chr.17:37406877-37479725 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TADA2A | NM_001166105.2 | NP_001159577.2 | ||
| NM_001291918.1 | NP_001278847.1 | |||
| NM_001488.4 | NP_001479.4 | |||
| NM_133439.3 | NP_597683.3 | |||
| XM_006722044.3 | XP_006722107.1 | |||
| XM_011525153.2 | XP_011523455.1 | |||
| XM_011525154.2 | XP_011523456.1 | |||
| XM_011525155.2 | XP_011523457.1 | |||
| XM_017024981.1 | XP_016880470.1 | |||
| XM_017024982.1 | XP_016880471.1 | |||
| XM_017024983.1 | XP_016880472.1 | |||
| XM_017024984.1 | XP_016880473.1 | |||
| XM_017024985.1 | XP_016880474.1 | |||
| XM_017024986.1 | XP_016880475.1 | |||
| AF064094.1 | AAC26659.1 | |||
| AK022767.1 | ||||
| AK096371.1 | ||||
| AK225918.1 | ||||
| AL526501.3 | ||||
| BC001172.1 | AAH01172.1 | |||
| BC011753.2 | AAH11753.1 | |||
| CD101741.1 | ||||
| DA244842.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3640494 | Chr.17:37406555 - 37610947 on Build GRCh38 | Gain |
|
| dgv3157n100 | Chr.17:37219334 - 37462210 on Build GRCh38 | Gain |
|
| nsv1061860 | Chr.17:37403775 - 37469800 on Build GRCh38 | Gain |
|
| esv2758687 | Chr.17:37211817 - 37414232 on Build GRCh38 | Gain |
|
| esv3640493 | Chr.17:37399339 - 37415811 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map