Assay Details
Target Gene Details
Entrez Gene ID: | 9054 |
Gene Name: | NFS1 cysteine desulfurase |
Gene Aliases: |
HUSSY-08, IscS, NIFS |
Location: |
Chr.20:35668688-35699365 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFS1 | NM_001198989.1 | NP_001185918.1 | ||
| NM_021100.4 | NP_066923.3 | |||
| NR_037570.2 | ||||
| AF097025.2 | AAD09187.2 | |||
| AJ010952.1 | CAA09424.1 | |||
| AK001265.1 | ||||
| AK001470.1 | ||||
| AK056242.1 | ||||
| AK091049.1 | ||||
| AK223239.1 | BAD96959.1 | |||
| AK295215.1 | ||||
| AK297969.1 | ||||
| AK301969.1 | ||||
| AK302023.1 | ||||
| AK312293.1 | ||||
| BC065560.1 | AAH65560.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526188 | Chr.20:35576541 - 36019079 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map