Assay Details
Target Gene Details
Entrez Gene ID: | 64785 |
Gene Name: | GINS complex subunit 3 |
Gene Aliases: |
PSF3 |
Location: |
Chr.16:58392394-58406144 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GINS3 | NM_001126129.1 | 3 | 582 | NP_001119601.1 |
| NM_001126130.1 | NP_001119602.1 | |||
| NM_022770.3 | 2 | 465 | NP_073607.2 | |
| AK023974.1 | BAB14747.1 | |||
| AK127454.1 | 1 | 227 | ||
| AK315622.1 | 2 | 416 | ||
| BC005879.1 | 2 | 340 | AAH05879.1 | |
| BC014437.1 | 2 | 359 | AAH14437.1 | |
| BP374281.1 | 3 | 525 | ||
| CR457283.1 | CAG33564.1 | |||
| DA550725.1 | 2 | 465 | ||
| KU178752.1 | 2 | 257 | ||
| KU178753.1 | 3 | 374 | ||
| KU178754.1 | 3 | 316 | ||
| KU178755.1 |
More Information
Set Membership: |
|

Genomic Map