Assay Details
Target Gene Details
Entrez Gene ID: | 594855 |
Gene Name: | complexin 3 |
Gene Aliases: |
CPX-III, CPXIII, Nbla11589 |
Location: |
Chr.15:74826610-74831795 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPLX3 | NM_001030005.2 | NP_001025176.1 | ||
| AB072900.1 | BAE45711.1 | |||
| AK024055.1 | BAB14805.1 | |||
| AY286501.1 | AAP41127.1 | |||
| BC018026.1 | AAH18026.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520527 | Chr.15:74816295 - 74830515 on Build GRCh38 | Loss |
|
| nsv833055 | Chr.15:74696003 - 74873947 on Build GRCh38 | Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map