Assay Details
Target Gene Details
Entrez Gene ID: | 256356 |
Gene Name: | glycerol kinase 5 (putative) |
Gene Aliases: |
- |
Location: |
Chr.3:142157527-142225607 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GK5 | NM_001039547.2 | 16 | 2824 | NP_001034636.1 |
| NR_033289.1 | 17 | 3907 | ||
| XM_011512624.2 | 16 | 2622 | XP_011510926.1 | |
| XM_017006089.1 | 17 | 2887 | XP_016861578.1 | |
| XM_017006090.1 | 15 | 2735 | XP_016861579.1 | |
| AK090901.1 | 7 | 1873 | BAC03542.1 | |
| BX648681.1 | 17 | 3844 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751979 | Chr.3:142135084 - 142296748 on Build GRCh38 | Gain |
|
| esv2763767 | Chr.3:142102103 - 142382712 on Build GRCh38 | Gain |
|
| esv3597995 | Chr.3:142099424 - 142172727 on Build GRCh38 | Gain |
|
| dgv8625n54 | Chr.3:142093917 - 142265796 on Build GRCh38 | Gain |
|
| dgv4912n100 | Chr.3:142100008 - 142372619 on Build GRCh38 | Gain |
|
| esv3597996 | Chr.3:142099805 - 142311818 on Build GRCh38 | Gain |
|
| dgv619n27 | Chr.3:142103779 - 142353090 on Build GRCh38 | Gain |
|
| nsv1008873 | Chr.3:142102091 - 142275700 on Build GRCh38 | Gain |
|
| esv2759182 | Chr.3:142010028 - 142410994 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map