Assay Details
Target Gene Details
Entrez Gene ID: | 1264 |
Gene Name: | calponin 1 |
Gene Aliases: |
HEL-S-14, SMCC, Sm-Calp |
Location: |
Chr.19:11538717-11550323 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNN1 | NM_001299.5 | 7 | 1079 | NP_001290.2 |
| NM_001308341.1 | 8 | 1354 | NP_001295270.1 | |
| NM_001308342.1 | 8 | 1345 | NP_001295271.1 | |
| XM_005259741.2 | 7 | 928 | XP_005259798.1 | |
| XM_017026289.1 | 7 | 1251 | XP_016881778.1 | |
| AK223234.1 | 7 | 945 | BAD96954.1 | |
| AK300837.1 | 7 | 859 | ||
| AK301888.1 | 8 | 1218 | ||
| AK313255.1 | ||||
| AK316406.1 | 8 | 1295 | ||
| BC022015.2 | 7 | 1032 | AAH22015.1 | |
| BC036307.1 | 7 | 1018 | AAH36307.2 | |
| D17408.1 | 7 | 960 | BAA04231.1 | |
| D86058.1 | BAA12983.1 | |||
| EU794601.1 | 7 | 1018 | ||
| S80560.1 | 7 | 942 | AAB35751.1 | |
| U37019.1 | 7 | 946 | AAC51780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509721 | Chr.19:11525216 - 11691390 on Build GRCh38 | Insertion |
|
| nsv519802 | Chr.19:11236817 - 11578343 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map