Assay Details
Target Gene Details
Entrez Gene ID: | 7786 |
Gene Name: | mitogen-activated protein kinase kinase kinase 12 |
Gene Aliases: |
DLK, MEKK12, MUK, ZPK, ZPKP1 |
Location: |
Chr.12:53480492-53501066 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K12 | NM_001193511.1 | 13 | 2629 | NP_001180440.1 |
| NM_006301.3 | 14 | 2530 | NP_006292.3 | |
| XM_005269138.3 | 13 | 2557 | XP_005269195.1 | |
| XM_006719588.3 | 13 | 2451 | XP_006719651.1 | |
| XM_011538725.2 | 13 | 3434 | XP_011537027.1 | |
| XM_017019956.1 | 14 | 2458 | XP_016875445.1 | |
| AB209453.1 | 5 | 3954 | BAD92690.1 | |
| AK094195.1 | 13 | 2425 | ||
| AK314530.1 | 13 | 2788 | ||
| BC050050.1 | AAH50050.1 | |||
| U07358.1 | 14 | 2357 | AAA67343.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv714 | Chr.12:53455169 - 53489159 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map