Assay Details
Target Gene Details
Entrez Gene ID: | 51283 |
Gene Name: | bifunctional apoptosis regulator |
Gene Aliases: |
BAR, RNF47 |
Location: |
Chr.16:14632811-14669236 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BFAR | NM_016561.2 | NP_057645.1 | ||
| XM_005255350.2 | XP_005255407.1 | |||
| XM_011522520.1 | XP_011520822.1 | |||
| AF173003.1 | AAF59975.1 | |||
| AK291114.1 | ||||
| AK297067.1 | ||||
| AK300778.1 | ||||
| AK313890.1 | ||||
| BC003054.1 | AAH03054.1 | |||
| BI912683.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833144 | Chr.16:14587299 - 14813650 on Build GRCh38 | Gain |
|
| dgv2738n100 | Chr.16:14617525 - 15031584 on Build GRCh38 | Gain |
|
| esv2758632 | Chr.16:14528198 - 15272693 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map