Assay Details
Target Gene Details
Entrez Gene ID: | 57546 |
Gene Name: | pyruvate dehyrogenase phosphatase catalytic subunit 2 |
Gene Aliases: |
PPM2B, PPM2C2 |
Location: |
Chr.16:66880397-66891101 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDP2 | NM_020786.2 | 2 | 1869 | NP_065837.1 |
| XM_005256064.4 | 2 | 1916 | XP_005256121.1 | |
| XM_005256066.4 | 2 | 1958 | XP_005256123.1 | |
| XM_005256067.4 | 2 | 1983 | XP_005256124.1 | |
| XM_005256068.4 | 3 | 1930 | XP_005256125.1 | |
| XM_017023506.1 | 3 | 2064 | XP_016878995.1 | |
| AB037769.1 | 2 | 1816 | ||
| AK292539.1 | 2 | 1869 | ||
| BC028030.1 | 2 | 1804 | AAH28030.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv572904 | Chr.16:66876182 - 66938470 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map