Assay Details
Target Gene Details
Entrez Gene ID: | 550631 |
Gene Name: | coiled-coil domain containing 157 |
Gene Aliases: |
- |
Location: |
Chr.22:30356635-30378655 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC157 | NM_001017437.3 | NP_001017437.2 | ||
| NM_001318334.1 | NP_001305263.1 | |||
| XM_006724278.3 | XP_006724341.1 | |||
| XM_006724279.3 | XP_006724342.1 | |||
| XM_006724280.3 | XP_006724343.1 | |||
| XM_011530263.2 | XP_011528565.1 | |||
| XM_011530266.2 | XP_011528568.1 | |||
| XM_011530267.2 | XP_011528569.1 | |||
| XM_011530268.2 | XP_011528570.1 | |||
| XM_011530269.2 | XP_011528571.1 | |||
| XM_011530270.2 | XP_011528572.1 | |||
| BC092419.1 | AAH92419.1 | |||
| CR936833.1 |
Target Gene Details
Entrez Gene ID: | 85371 |
Gene Name: | KIAA1656 protein |
Gene Aliases: |
- |
Location: |
Chr.22:30368811-30377905 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA1656 | NR_046312.1 | 2 | 4718 | |
| AB051443.1 | 1 | 4086 | BAB33326.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953067 | Chr.22:30365112 - 30395011 on Build GRCh38 | Deletion |
|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|

Genomic Map