Assay Details
Target Gene Details
Entrez Gene ID: | 2353 |
Gene Name: | Fos proto-oncogene, AP-1 transcription factor subunit |
Gene Aliases: |
AP-1, C-FOS, p55 |
Location: |
Chr.14:75278778-75282234 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOS | NM_005252.3 | 4 | 1070 | NP_005243.1 |
| AK097379.1 | 4 | 835 | ||
| AK290907.1 | 3 | 958 | ||
| AK291326.1 | 4 | 1020 | ||
| AK298659.1 | 3 | 912 | ||
| BC004490.2 | 4 | 991 | AAH04490.1 | |
| BX647104.1 | 4 | 1020 | ||
| CR541785.1 | 4 | 865 | CAG46584.1 | |
| CR542267.1 | 4 | 865 | CAG47063.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832833 | Chr.14:75100965 - 75304860 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map