Assay Details
Target Gene Details
Entrez Gene ID: | 4776 |
Gene Name: | nuclear factor of activated T-cells 4 |
Gene Aliases: |
NF-AT3, NF-ATC4, NFAT3 |
Location: |
Chr.14:24366911-24379604 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952819 | Chr.14:24367695 - 24377694 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map