Assay Details
Target Gene Details
Entrez Gene ID: | 6641 |
Gene Name: | syntrophin beta 1 |
Gene Aliases: |
59-DAP, A1B, BSYN2, DAPA1B, SNT2, SNT2B1, TIP-43 |
Location: |
Chr.8:120535745-120812069 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNTB1 | NM_021021.3 | 7 | 1878 | NP_066301.1 |
| AK026095.1 | 9 | 2021 | ||
| AK292655.1 | 7 | 1952 | ||
| BC098573.1 | 7 | 1838 | AAH98573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519152 | Chr.8:120446217 - 120559026 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map