Assay Details
Target Gene Details
Entrez Gene ID: | 5009 |
Gene Name: | ornithine carbamoyltransferase |
Gene Aliases: |
OCTD |
Location: |
Chr.X:38352483-38421450 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OTC | NM_000531.5 | NP_000522.3 | ||
| BC107153.1 | AAI07154.1 | |||
| BC107154.1 | AAI07155.1 | |||
| BC114496.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573871 | Chr.X:38347596 - 38352990 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map