Assay Details
Target Gene Details
Entrez Gene ID: | 164633 |
Gene Name: | calcium binding protein 7 |
Gene Aliases: |
CALN2 |
Location: |
Chr.22:29720090-29731833 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CABP7 | NM_182527.2 | NP_872333.1 | ||
| XM_006724161.3 | XP_006724224.1 | |||
| BC051805.1 | AAH51805.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471194 | Chr.22:29707487 - 29732391 on Build GRCh38 | Gain |
|
| nsv515720 | Chr.22:29685726 - 29767537 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map