Assay Details
Target Gene Details
Entrez Gene ID: | 7528 |
Gene Name: | YY1 transcription factor |
Gene Aliases: |
DELTA, INO80S, NF-E1, UCRBP, YIN-YANG-1 |
Location: |
Chr.14:100238765-100279034 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YY1 | NM_003403.4 | 1 | 127 | NP_003394.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3895n54 | Chr.14:100235080 - 100241246 on Build GRCh38 | Gain |
|
| esv3635496 | Chr.14:100169585 - 100290965 on Build GRCh38 | Gain |
|
| dgv3896n54 | Chr.14:100237676 - 100241246 on Build GRCh38 | Loss |
|
| esv22463 | Chr.14:100238164 - 100240686 on Build GRCh38 | Loss |
|
| nsv827052 | Chr.14:100237465 - 100241726 on Build GRCh38 | Gain |
|
| dgv3897n54 | Chr.14:100238205 - 100240715 on Build GRCh38 | Gain |
|
| nsv832875 | Chr.14:100192592 - 100368905 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map