Assay Details
Target Gene Details
Entrez Gene ID: | 6232 |
Gene Name: | ribosomal protein S27 |
Gene Aliases: |
MPS-1, MPS1, S27 |
Location: |
Chr.1:153990763-153992155 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS27 | AK312070.1 | |||
| CD177656.1 | ||||
| D28454.1 | BAA05820.1 | |||
| L19739.1 | AAA59867.1 | |||
| U57847.1 | AAB02266.1 |
More Information
Set Membership: |
|

Genomic Map