Assay Details
Target Gene Details
Entrez Gene ID: | 11021 |
Gene Name: | RAB35, member RAS oncogene family |
Gene Aliases: |
H-ray, RAB1C, RAY |
Location: |
Chr.12:120095094-120116839 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB35 | NM_001167606.1 | NP_001161078.1 | ||
| NM_006861.6 | NP_006852.1 | |||
| XM_011537779.1 | XP_011536081.1 | |||
| XM_017018723.1 | XP_016874212.1 | |||
| AF498960.1 | AAM21108.1 | |||
| AK125543.1 | ||||
| AK300127.1 | ||||
| AK304620.1 | ||||
| AK312538.1 | ||||
| BC015931.2 | AAH15931.1 | |||
| BM467132.1 | ||||
| BQ233709.1 | ||||
| BT020024.1 | AAV38827.1 | |||
| CR536486.1 | CAG38725.1 | |||
| CR541683.1 | CAG46484.1 | |||
| DC349830.1 | ||||
| DC370610.1 | ||||
| DC408044.1 | ||||
| X79781.1 | CAA56177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952492 | Chr.12:120090097 - 120106496 on Build GRCh38 | Deletion |
|
| nsv560433 | Chr.12:120095892 - 120238287 on Build GRCh38 | Loss |
|
| nsv560434 | Chr.12:120096343 - 120107914 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map