Assay Details
Target Gene Details
Entrez Gene ID: | 125965 |
Gene Name: | cytochrome c oxidase subunit 6B2 |
Gene Aliases: |
COXVIB2, CT59 |
Location: |
Chr.19:55349702-55354814 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX6B2 | NM_144613.4 | 4 | 464 | NP_653214.2 |
| AK057427.1 | 4 | 361 | ||
| AY152398.1 | 4 | 370 | AAN46751.1 | |
| BC026123.2 | 4 | 465 | ||
| BC100899.2 | 3 | 301 | AAI00900.1 | |
| BC100900.1 | 3 | 301 | AAI00901.1 | |
| BC100901.1 | 3 | 301 | AAI00902.1 | |
| BC100902.1 | 3 | 301 | AAI00903.1 | |
| BG717038.1 | 4 | 469 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953624 | Chr.19:55302033 - 55625034 on Build GRCh38 | Deletion |
|
| nsv470162 | Chr.19:55334018 - 55390889 on Build GRCh38 | Loss |
|
| nsv516199 | Chr.19:55342945 - 55389731 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map